Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XTerra Shield RP18 Guard Cartridge, 10 μm, 30x10 mm

Catalog No. 502390733 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

  • Designed to protect HPLC and SFC 30 mm ID preparative columns.
  • Extends column lifetimes and reduces purification costs.
  • Enhances analytical performance by lowering particle contamination.
  • Compatible with 30 mm I.D. analytical columns with a 10 μm sorbent particle size.
  • Can sustain high pressures in various applications.
  • Undergoes quality control testing using a peptide map for consistent performance.
  • Works well across a broad pH range of 2 to 12.
  • Withstands a maximum temperature of 60°C.

Catalog No. 50-239-0733 Supplier Waters Corp Supplier No. 186006905
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Designed to protect HPLC and SFC 30 mm ID preparative columns.
  • Extends column lifetimes and reduces purification costs.
  • Enhances analytical performance by lowering particle contamination.
  • Compatible with 30 mm I.D. analytical columns with a 10 μm sorbent particle size.
  • Can sustain high pressures in various applications.
  • Undergoes quality control testing using a peptide map for consistent performance.
  • Works well across a broad pH range of 2 to 12.
  • Withstands a maximum temperature of 60°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.