Learn More
Waters Corp VanGuard Cartridge, Shield RP18, 5 μm, 125 Å, 2.1 mm × 5 mm
Supplier:  Waters Corp 186007933
Encompass_Preferred
XTerra Shield RP18 VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm ID analytical columns containing 5 μm sorbent particles
  • Features C18 chemistry for reversed phase separation
  • Constructed with hybrid particle substrate
  • Maintains performance across a pH range of 2 to 12
  • Withstands maximum pressure of 18000 psi (1240 Bar)
  • Endcapped for enhanced stability
  • Low silanol activity for reduced secondary interactions
  • Spherical particle shape for consistent flow
  • Pore size of 125 Å
  • Suitable for HPLC systems
  • USP classification L1
Catalog No. 50-239-2462