Learn More
Waters Corp XTerra Shield RP18 VanGuard Cartridge, 3.5 μm, 2.1 mm x 5 mm
Catalog No. 502392664 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XTerra Shield RP18 VanGuard Cartridges extend the lifetime and enhance the performance of analytical columns. They achieve this by filtering out particulate contamination from the mobile phase stream. This cartridge is optimized for protecting 2.1 mm and 3.0 mm I.D. analytical columns that contain 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination.
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns.
  • Designed for 3.5 μm sorbent particles.
Catalog No. 50-239-2664 Supplier Waters Corp Supplier No. 186007929
XTerra Shield RP18 VanGuard Cartridges extend the lifetime and enhance the performance of analytical columns. They achieve this by filtering out particulate contamination from the mobile phase stream. This cartridge is optimized for protecting 2.1 mm and 3.0 mm I.D. analytical columns that contain 3.5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination.
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns.
  • Designed for 3.5 μm sorbent particles.