Learn More
Waters Corp XTerra Shield RP8 Column, 125 Å, 5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186000493RF
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled device-tagged columns were developed for automatic identification, tracking, and digital tracing of column use history. eConnect columns can be used on any HPLC, but their advantages are unlocked when used with a compatible iS System running Empower software.
  • Equipped with an electronic eConnect column tag containing unique column-specific information.
  • NFC-enabled device-tagged columns developed for automatic identification, tracking, and digital tracing of column use history.
  • Can be used on any HPLC.
  • Advantages are unlocked when used with a compatible iS System running Empower software.
Catalog No. 50-239-2213