Learn More
Waters Corp HPLC Column, 125Å Pore, 3.5 μm Particle, 4.6 mm x 150 mm
Catalog No. 502392945 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra Shield RP8 Column is an eConnect HPLC column. It features second-generation smart-column technology with a permanently attached electronic tag for storing product information. This tag enables NFC access to product details and allows data to be automatically imported and searched through HPLC system software.
  • Pre-equipped eConnect Tag simplifies setup and reduces manual data entry.
  • Permanent, tamper-proof tag reduces compliance concerns and simplifies system setup.
  • Fit-for-purpose tag design improves detection and mitigates interference.
  • NFC-capable tag provides convenient access to column information.
  • Equivalent to untagged counterparts in quality and performance.
Catalog No. 50-239-2945 Supplier Waters Corp Supplier No. 186000443RF
The XTerra Shield RP8 Column is an eConnect HPLC column. It features second-generation smart-column technology with a permanently attached electronic tag for storing product information. This tag enables NFC access to product details and allows data to be automatically imported and searched through HPLC system software.
  • Pre-equipped eConnect Tag simplifies setup and reduces manual data entry.
  • Permanent, tamper-proof tag reduces compliance concerns and simplifies system setup.
  • Fit-for-purpose tag design improves detection and mitigates interference.
  • NFC-capable tag provides convenient access to column information.
  • Equivalent to untagged counterparts in quality and performance.