Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XTerra OBD Prep Column, 125Å, 10μm, 10 mm, 300 mm

Supplier:  Waters Corp 186008140

Encompass_Preferred

XTerra Preparative RP8 columns are hybrid-based, reversed-phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first-generation columns provide superior pH stability compared to a silica-based column.

  • C8 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 2 to 12
  • Temperature limits up to 60°C
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Shield RP8 bonding technology
  • Low silanol activity
  • Spherical particle shape
  • OBD prep column format
  • Surface area of 175
  • HPLC preparative system
  • LC/MS and LC techniques
  • L7 USP classification
  • 14% carbon load
  • 1 unit per package

Catalog No. 50-239-2358


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.