Learn More
Waters Corp Prep Column, XTerra Shield RP8 OBD, 125Å, 10 μm, 10 mm x 150 mm
Catalog No. 502392553 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XTerra Shield RP8 OBD Prep Column utilizes hybrid particle technology for superior chromatographic performance. It features methylene bridged particles and an embedded polar group, ensuring optimal peak shape, unique selectivity for basic compounds, and excellent pH stability.
  • Hybrid particle technology offers efficiency and stability.
  • Methylene bridged particles enhance performance.
  • Embedded polar group delivers optimal peak shape and unique selectivity.
  • Shield technology ensures excellent peak shapes for bases.
  • Wide pH range (2–12) increases selectivity and improves resolution at high pH.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60x) versus conventional silica.
Catalog No. 50-239-2553 Supplier Waters Corp Supplier No. 186008132
Add to Cart
Add to Cart
The XTerra Shield RP8 OBD Prep Column utilizes hybrid particle technology for superior chromatographic performance. It features methylene bridged particles and an embedded polar group, ensuring optimal peak shape, unique selectivity for basic compounds, and excellent pH stability.
  • Hybrid particle technology offers efficiency and stability.
  • Methylene bridged particles enhance performance.
  • Embedded polar group delivers optimal peak shape and unique selectivity.
  • Shield technology ensures excellent peak shapes for bases.
  • Wide pH range (2–12) increases selectivity and improves resolution at high pH.
  • Excellent water wettability, even in 100% aqueous mobile phases.
  • Easy scale-up from analytical to preparative chromatography.
  • Increased mass loading (up to 60x) versus conventional silica.