Learn More
Waters Corp XTerra VanGuard Cartridge, 125Å, 5 μm, 2.1 mm × 5 mm
Supplier:  Waters Corp 186007945
Encompass_Preferred
XTerra Shield RP8 VanGuard Cartridges are used to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. XTerra Shield RP8 analytical columns containing 5 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. analytical columns containing 5 μm sorbent particles.
Catalog No. 50-239-1768