Learn More
Waters Corp VanGuard Cartridge, XTerra Shield RP8, 125Å, 3.5 μm
Catalog No. 502391383 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge protects analytical columns containing 3.5 μm sorbent particles and is easy to set up and use. It shields columns against contaminants, increasing and improving analytical performance.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns.
  • Shields columns against chemical and particle impurities.
  • Increases and improves the analytical performance of the column.
  • Designed for routine use and sustains high pressures in UPLC applications.
  • Each batch is QC tested utilizing a peptide map for consistent performance.
  • Designed for use with columns in reversed-phase liquid chromatography.
  • Quickly mounts to the intake side of a column.
  • Operates across a wide pH range of 2 to 12 pH.
Catalog No. 50-239-1383 Supplier Waters Corp Supplier No. 186007943
VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge protects analytical columns containing 3.5 μm sorbent particles and is easy to set up and use. It shields columns against contaminants, increasing and improving analytical performance.
  • Optimized to protect 3.9 mm and 4.6 mm I.D. analytical columns.
  • Shields columns against chemical and particle impurities.
  • Increases and improves the analytical performance of the column.
  • Designed for routine use and sustains high pressures in UPLC applications.
  • Each batch is QC tested utilizing a peptide map for consistent performance.
  • Designed for use with columns in reversed-phase liquid chromatography.
  • Quickly mounts to the intake side of a column.
  • Operates across a wide pH range of 2 to 12 pH.