Learn More
Aviva Systems Biology Corp XTP3TPA Antibody - middle region (ARP46329_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46329_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against XTP3TPA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human XTP3TPA. Alternate names: CDA03, MGC5627, RS21C6, XTP3TPAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human
Catalog No. 50-014-59304