Learn More
Sino Biological XTP3TPA / DCTPP1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 14577R074100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human XTP3TPA / DCTPP1 (rh XTP3TPA / DCTPP1; Catalog#14577-H07E; NP_077001.1; Met1-Thr170).
Catalog No. 50-224-7924