Learn More
New Era Enterprises XTRA LNG TIP PPT NMR TBS 100CS
Catalog No. 50200129
Encompass_Preferred
Pipet Tip, New Era, Extra Long, For Use With 5mm x 8-9 in. Thin-walled NMR Tubes, 9.25 in. - TIP, 100 per box.
Catalog No. 50-200-129 Supplier New Era Enterprises Supplier No. NE201A
Pipet Tip, New Era, Extra Long, For Use With 5mm x 8-9 in. Thin-walled NMR Tubes, 9.25 in. - TIP, 100 per box.