Learn More
Alkali Scientific Xtreme™ 2 High-Strength Sealing Film, Large, 100/pk
SDP
Catalog No. 502388953
Encompass_Preferred
Xtreme™ 2 High-Strength Sealing Film an extra-strong thick adhesive to minimize evaporation, yet removes easily without leaving residue on the plate. Features chamfered corners to eliminate plate overhang for robotics applications and tight storage conditions. Certified DNase- & RNase- free. Ideal for use with raised-rim and non-skirted PCR plates.
Catalog No. 50-238-8953 Supplier Alkali Scientific Supplier No. SF10MLXL
Xtreme™ 2 High-Strength Sealing Film an extra-strong thick adhesive to minimize evaporation, yet removes easily without leaving residue on the plate. Features chamfered corners to eliminate plate overhang for robotics applications and tight storage conditions. Certified DNase- & RNase- free. Ideal for use with raised-rim and non-skirted PCR plates.