Learn More
Alkali Scientific Xtreme™ AlumaFoil™ High-Strength ULT Sealing Film, 100/pk
SDP
Catalog No. 502389254
Encompass_Preferred
Xtreme™ AlumaFoil™ is an aluminum foil seal that offers excellent chemical resistance and superior performance in ultra-low temperatures. The smaller dimensions and chamfered corners result in no plate overhang for robotics applications; universal fit for use with all plate types (e.g. raised-rim and non-skirted). Certified DNase- & RNase- free. Recommended for -80°C long-term storage. Excellent chemical resistance (e.g. DMSO). Specially formulated adhesive provides an excellent seal to minimize evaporation and well-to-well contamination. 1.9mil thick aluminum foil removes easily without tearing and allows for visual verification of the seal’s security around each well. Pierceable with a pipet tip for direct sample recovery. Not recommended for PCR.
Catalog No. 50-238-9254 Supplier Alkali Scientific Supplier No. SF11ML
Xtreme™ AlumaFoil™ is an aluminum foil seal that offers excellent chemical resistance and superior performance in ultra-low temperatures. The smaller dimensions and chamfered corners result in no plate overhang for robotics applications; universal fit for use with all plate types (e.g. raised-rim and non-skirted). Certified DNase- & RNase- free. Recommended for -80°C long-term storage. Excellent chemical resistance (e.g. DMSO). Specially formulated adhesive provides an excellent seal to minimize evaporation and well-to-well contamination. 1.9mil thick aluminum foil removes easily without tearing and allows for visual verification of the seal’s security around each well. Pierceable with a pipet tip for direct sample recovery. Not recommended for PCR.