Learn More
Lonza Walkersville XTREME CHO FEED Powder 10L
Catalog No. 5000496204
Encompass
XTREME CHO Feed Powder is a chemically defined protein-free and non-animal origin basal powder feed to produce 10L of liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media PowerCHO 2 The XTREME CHO Feed Powder has low salts for better osmolarity control This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS
Catalog No. 50-004-96204 Supplier Lonza Walkersville Supplier No. VPW098D
XTREME CHO Feed Powder is a chemically defined protein-free and non-animal origin basal powder feed to produce 10L of liquid feed The feed is formulated to maximize cell growth using Lonza CHO cell culture media PowerCHO 2 The XTREME CHO Feed Powder has low salts for better osmolarity control This BioWhittaker Cell Culture Media is produced according to cGMP and labelled For Further Manufacturing (FFM) GMP Products are intended for research or further manufacturing use only THIS PRODUCT IS NOT INTENDED FOR DIRECT THERAPEUTIC USE IN HUMANS