Learn More
Boca Scientific Inc Xtreme DNA Purification Kit for Buccal Swabs and Saliva, 50 reactions
Catalog No. NC0706231
Encompass
The Xtreme DNA Isolation Kit is a silica membrane-based spin column DNA purification kit designed to isolate genomic DNA from buccal swabs and saliva with minimal losses. The isolated DNA is highly purified with a high molecular weight and is free of trace PCR inhibitors. The DNA is suitable for use in even the most demanding downstream applications. This kit does not contain solvent-based chemicals.
Catalog No. NC0706231 Supplier Boca Scientific Inc Supplier No. XME50
The Xtreme DNA Isolation Kit is a silica membrane-based spin column DNA purification kit designed to isolate genomic DNA from buccal swabs and saliva with minimal losses. The isolated DNA is highly purified with a high molecular weight and is free of trace PCR inhibitors. The DNA is suitable for use in even the most demanding downstream applications. This kit does not contain solvent-based chemicals.