Learn More
Alkali Scientific Xtreme™ High-Strength Sealing Film, 100/pk
SDP
Catalog No. 502388692
Encompass_Preferred
Xtreme™ Seal features an extra-strong thick adhesive to minimize evaporation, yet removes easily without leaving residue on the plate. Features chamfered corners to eliminate plate overhang for robotics applications and tight storage conditions. Certified DNase- & RNase- free. Ideal for use with raised-rim and non-skirted PCR plates. Selection of the proper film size will minimize evaporation. The film should not overhang or ride up the wall of a raised-rim plate. More complete coverage of skirted flat-top plates will reduce evaporation in the outer wells. End-tab removal before thermal cycling is highly recommended.
Catalog No. 50-238-8692 Supplier Alkali Scientific Supplier No. SF12ML
Xtreme™ Seal features an extra-strong thick adhesive to minimize evaporation, yet removes easily without leaving residue on the plate. Features chamfered corners to eliminate plate overhang for robotics applications and tight storage conditions. Certified DNase- & RNase- free. Ideal for use with raised-rim and non-skirted PCR plates. Selection of the proper film size will minimize evaporation. The film should not overhang or ride up the wall of a raised-rim plate. More complete coverage of skirted flat-top plates will reduce evaporation in the outer wells. End-tab removal before thermal cycling is highly recommended.