Learn More
Shelby™ Xtrication™ Gloves
Catalog No. 19821453 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
X-Large
Catalog No. Size
19-821-453 X-Large
1 options
Catalog No. 19-821-453 Supplier Shelby™ Supplier No. 2500XL
Anatomically designed for improved hand and finger dexterity
  • Glove body is constructed of durable Nylon Cordura™
  • Palm and finger reinforcements cover all high-wear stress points with Aramid Fiber fabric
  • Rubberized fingers enhance grip in wet or slippery situations
  • Black with chartreuse glove back for improved visibility
  • Shirred elastic wrist closure
Specifications
Certifications/Compliance CE PPE, EN 659:1996, EN 388:1994
Color Black
Material Cordura Nylon
Product Type Extrication Gloves
Size X-Large
Liner Material Fluffed Terry Cotton