Learn More
Cayman Chemical XTT (sodium salt hydrate), Purity: ≥98%, CAS: 413585-64-5, 100 mg
SDP
Catalog No. 502968508
Encompass_Preferred
A cell-impermeable, negatively charged tetrazolium dye that produces a water-soluble formazan when reduced at the cell surface by cellular-derived NADH and an electron mediator
Catalog No. 50-296-8508 Supplier Cayman Chemical Supplier No. 14919100 MG
A cell-impermeable, negatively charged tetrazolium dye that produces a water-soluble formazan when reduced at the cell surface by cellular-derived NADH and an electron mediator