Learn More
Cayman Chemical XTT (sodium salt hydrate), Purity: ≥98%, CAS: 413585-64-5, 50 mg
SDP
Catalog No. 502971564
Encompass_Preferred
A cell-impermeable, negatively charged tetrazolium dye that produces a water-soluble formazan when reduced at the cell surface by cellular-derived NADH and an electron mediator
Catalog No. 50-297-1564 Supplier Cayman Chemical Supplier No. 1491950 MG
A cell-impermeable, negatively charged tetrazolium dye that produces a water-soluble formazan when reduced at the cell surface by cellular-derived NADH and an electron mediator