Learn More
Crescent Chemical Co Inc XTT
SDP
Supplier:  Crescent Chemical Co Inc SE38450.01
Encompass_Preferred
XTT Monotetrazolium salt which forms a water-soluble formazan upon reduction. Suitable for anti-HIV (1 - 3) and anti-tumor (4 - 6) drug testing as well as for cell proliferation assays (7 - 8). Size - 50MG Storage Conditions - +2 °C TO +8 °C
Catalog No. 50-248-298