Learn More
Apexbio Technology LLC XTT CELL PROLIF KIT-1000mg
SDP
Catalog No. 5000897470
Encompass_Preferred
The XTT Cell Proliferation Assay Kit is a crucial tool in biomedical research used to assess cell viability and proliferation in various studies Originating from colorimetric assays it measures the activity of mitochondrial enzymes that reduce the tetrazolium compound XTT to a colored formazan product indicating metabolically active cells This assay is commonly used in in vitro settings for drug screening and evaluating cytotoxicity in cell lines It shows activity in the nanomolar to low micromolar range depending on cell type and conditions Typical applications include testing cancer cell models evaluating potential therapeutic agents and conducting high-throughput screening due to its non-radioactive and sensitive nature Experimental concentrations typically depend on the specific research design allowing for flexibility and precision in diverse experimental setups
Catalog No. 50-008-97470 Supplier Apexbio Technology LLC Supplier No. K22721000
The XTT Cell Proliferation Assay Kit is a crucial tool in biomedical research used to assess cell viability and proliferation in various studies Originating from colorimetric assays it measures the activity of mitochondrial enzymes that reduce the tetrazolium compound XTT to a colored formazan product indicating metabolically active cells This assay is commonly used in in vitro settings for drug screening and evaluating cytotoxicity in cell lines It shows activity in the nanomolar to low micromolar range depending on cell type and conditions Typical applications include testing cancer cell models evaluating potential therapeutic agents and conducting high-throughput screening due to its non-radioactive and sensitive nature Experimental concentrations typically depend on the specific research design allowing for flexibility and precision in diverse experimental setups