Learn More
Cayman Chemical XTT Cell Proliferation Assay Kit, 480 Well
SDP
Catalog No. 502886860
Encompass_Preferred
Cayman’s XTT Cell Proliferation Assay provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model. The assay is based on the extracellular reduction of XTT by NADH produced in the mitochondria via trans-plasma membrane electron transport and an electron mediator. Reduction of XTT produces a water-soluble formazan that dissolves directly into the culture medium, eliminating the need for an additional solubilization step. This kit will allow investigators to screen drug candidates involved in cell cycle regulation.
Catalog No. 50-288-6860 Supplier Cayman Chemical Supplier No. 10010200480 WELL
Cayman’s XTT Cell Proliferation Assay provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model. The assay is based on the extracellular reduction of XTT by NADH produced in the mitochondria via trans-plasma membrane electron transport and an electron mediator. Reduction of XTT produces a water-soluble formazan that dissolves directly into the culture medium, eliminating the need for an additional solubilization step. This kit will allow investigators to screen drug candidates involved in cell cycle regulation.