Learn More
TRIBIOSCIENCE INC XTT Cell Viability Assay, 500 Tests
SDP
Catalog No. NC3454573
Encompass_Preferred
Tribioscience’s XTT Cell Viability Assay Kit is a colorimetric assay to quantify the viable cells with soluble formazan dye. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.
Catalog No. NC3454573 Supplier TRIBIOSCIENCE INC Supplier No. TBS2021500
Tribioscience’s XTT Cell Viability Assay Kit is a colorimetric assay to quantify the viable cells with soluble formazan dye. The assay can be used for the measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds.