Learn More
Medchemexpress LLC XTT (sodium) | 111072-31-2 | 99.9% | 100 MG
SDP
Catalog No. 5000231068
Encompass_Preferred
XTT (sodium) is a tetrazolium-based reagent used to assess cell viability and cytotoxicity. Metabolically active cells reduce the water-soluble XTT sodium salt to an orange formazan, and the formazan absorbance is measured colorimetrically to quantify cell metabolic activity.
  • Produces a water-soluble orange formazan for direct colorimetric readout.
  • Measures metabolic activity of viable cells via tetrazolium reduction.
  • Suitable for mammalian cell viability and cytotoxicity assays.
  • Compatible with microplate and high-throughput formats.
  • Supplied as a sodium salt powder for preparation of aqueous working solutions.
Catalog No. 50-002-31068 Supplier Medchemexpress LLC Supplier No. HY122131100MG
XTT (sodium) is a tetrazolium-based reagent used to assess cell viability and cytotoxicity. Metabolically active cells reduce the water-soluble XTT sodium salt to an orange formazan, and the formazan absorbance is measured colorimetrically to quantify cell metabolic activity.
  • Produces a water-soluble orange formazan for direct colorimetric readout.
  • Measures metabolic activity of viable cells via tetrazolium reduction.
  • Suitable for mammalian cell viability and cytotoxicity assays.
  • Compatible with microplate and high-throughput formats.
  • Supplied as a sodium salt powder for preparation of aqueous working solutions.