Learn More
Gold Biotechnology Inc XTT Sodium Salt 100 mg
SDP
Catalog No. 5000562049
Encompass_Preferred
XTT sodium salt is used in the XTT colormetric assay and assists in identifying metabolic activity within cells In the XTT assay the metabolic activity of cells reduces XTT sodium salt to an orange color formazan dye by dehydrogenase enzymes Dye conversion will only occur in viable cells
Catalog No. 50-005-62049 Supplier Gold Biotechnology Inc Supplier No. X200100
XTT sodium salt is used in the XTT colormetric assay and assists in identifying metabolic activity within cells In the XTT assay the metabolic activity of cells reduces XTT sodium salt to an orange color formazan dye by dehydrogenase enzymes Dye conversion will only occur in viable cells