Learn More
Biospherix XVivo X3 Cell Incubation & Handling Platform
SDP
Catalog No. NC3622991
Encompass_Preferred
XVivo X3 Cell Incubation & Handling Platform - Independent Configuration - Single Air-lock & Gloved Processing Chamber w/ Internal Incubator Chamber - O2, CO2, Temperature, and HEPA controlled
Catalog No. NC3622991 Supplier Biospherix Supplier No. TFX3IND47
Add to Cart
Add to Cart
Request a Quote
XVivo X3 Cell Incubation & Handling Platform - Independent Configuration - Single Air-lock & Gloved Processing Chamber w/ Internal Incubator Chamber - O2, CO2, Temperature, and HEPA controlled