Learn More
Ampco XX-Small Valve Wheel Hook
SDP
Catalog No. 19175142 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
13 -3/4 in.
Catalog No. Length (English)
19-175-142 13 -3/4 in.
1 options
Catalog No. 19-175-142 Supplier Ampco Supplier No. WV32
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant valve wheel hook. Made in the USA.
  • F-type valve wheel hook
  • Aluminum bronze material, beryllium free
  • Includes lifetime warranty
  • Please see image for additional information regarding measurements
  • 13 and 3/4 in. length
  • a (jaw opening size): 1 and 3/8 in
  • b (throat depth): 2 and 3/8 in.
Specifications
Product Type Valve Wheel Lock
Length (English) 13 -3/4 in.
Quantity 1 Ea.