Learn More
TWD Scientific LLC Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3". 20/case
SDP
Catalog No. NC3502259 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3".Maximum Capacity (mL) >270mL
Catalog No. NC3502259 Supplier TWD Scientific LLC Supplier No. ASWF5XXLCS
Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3".Maximum Capacity (mL) >270mL