Learn More
TWD Scientific LLC Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3". 4/pack
SDP
Catalog No. NC3502257 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3"Maximum Capacity (mL) >270mL
Catalog No. NC3502257 Supplier TWD Scientific LLC Supplier No. ASWF5XXLPK
Disposable antistatic polypropylene weighing funnel, size XXL. Bottom Dimension: Approx. 3"x3"Maximum Capacity (mL) >270mL