Learn More
Electron Microscopy Sciences XX-Large Bulk Specimen Holder
SDP
Catalog No. 501927141
Encompass_Preferred
Machined aluminum specimen holder for scanning electron microscopy holds up to 3" specimens. Opening is 80 mm wide x 48 mm long x 13 mm deep. Stainless steel allen set screws keep specimen in place. 3.2 mm pin. Allen wrench included.
This item is not returnable. View return policy
Catalog No. 50-192-7141 Supplier Electron Microscopy Sciences Supplier No. 7595109
Machined aluminum specimen holder for scanning electron microscopy holds up to 3" specimens. Opening is 80 mm wide x 48 mm long x 13 mm deep. Stainless steel allen set screws keep specimen in place. 3.2 mm pin. Allen wrench included.