Learn More
BioSpec Products XXTuff™ non-sterile, reinforced 2 ml vial and cap, 500/pk
SDP
Supplier:  BioSpec Products 330TX
Encompass_Preferred
The XXTuff microvial has the same dimensions as a standard 2 ml microcentrifuge vial with screw cap and o-ring seal. However, it is molded from a special polypropylene resin and has been reinforced in critical areas. Unlike standard microvials, the XXTuff vial is able to withstand the rigors of dry grinding using steel beads*. Tests show that the XXTuff vial is over 10 times more durable than a standard microvial. 500/pk
Catalog No. NC0444131