Learn More
Medchemexpress LLC Xy-06-007 | 2757045-94-4 | 98.9% | 809.27 | C41H41ClN8O8 | 5 MG
SDP
Catalog No. 5000221458
Encompass_Preferred
XY-06-007 is a bump-and-hole (B&H) PROTAC designed to selectively degrade the BRD4BD1L94V variant. It demonstrates potent cellular activity (DC50, 6 h = 10 nM) and has reported pharmacokinetic properties suitable for in vivo research applications.
  • Selective bump-and-hole PROTAC for BRD4BD1L94V.
  • Potent cellular activity with a DC50 (6 h) of 10 nM.
  • High purity: 98.9% by HPLC.
  • Solid form; store powder at -20°C for long-term stability.
  • Suitable pharmacokinetic profile for in vivo studies.
  • Molecular formula C41H41ClN8O8; molecular weight 809.27.
Catalog No. 50-002-21458 Supplier Medchemexpress LLC Supplier No. HY1452265MG
XY-06-007 is a bump-and-hole (B&H) PROTAC designed to selectively degrade the BRD4BD1L94V variant. It demonstrates potent cellular activity (DC50, 6 h = 10 nM) and has reported pharmacokinetic properties suitable for in vivo research applications.
  • Selective bump-and-hole PROTAC for BRD4BD1L94V.
  • Potent cellular activity with a DC50 (6 h) of 10 nM.
  • High purity: 98.9% by HPLC.
  • Solid form; store powder at -20°C for long-term stability.
  • Suitable pharmacokinetic profile for in vivo studies.
  • Molecular formula C41H41ClN8O8; molecular weight 809.27.