Learn More
Medchemexpress LLC XY-06-007 | 2757045-94-4 | 98.9% | C41H41ClN8O8 | 1 MG
SDP
Catalog No. 5000355206
Encompass_Preferred
XY-06-007 is a selective and potent bump-and-hole (B&H)-PROTAC BRD4BD1L94V degrader that shows strong degradation activity against its target at nanomolar concentrations without affecting off-targets. It exhibits suitable pharmacokinetic properties for in vivo studies, making it a valuable tool for research applications.
  • Exhibits a half-degradation concentration (DC50, 6 h) of 10 nM against BRD4BD1L94V.
  • Demonstrates suitable pharmacokinetics for in vivo studies.
  • Has good plasma concentration, area under the curve (AUC), and bioavailability.
Catalog No. 50-003-55206 Supplier Medchemexpress LLC Supplier No. HY1452261MG
XY-06-007 is a selective and potent bump-and-hole (B&H)-PROTAC BRD4BD1L94V degrader that shows strong degradation activity against its target at nanomolar concentrations without affecting off-targets. It exhibits suitable pharmacokinetic properties for in vivo studies, making it a valuable tool for research applications.
  • Exhibits a half-degradation concentration (DC50, 6 h) of 10 nM against BRD4BD1L94V.
  • Demonstrates suitable pharmacokinetics for in vivo studies.
  • Has good plasma concentration, area under the curve (AUC), and bioavailability.