Learn More
Dino Lite MS15X-S2 XY/Rotation Base with C-Clamp - Designed for
Catalog No. NC1212662
Encompass_Preferred
MS15X-S1 is a multifunctional X/Y fine adjustment base designed for precise adjustment under Dino-Lite microscopes. The included rotating table connects directly to the top plate of the X/Y stage, allowing for simple rotation of the viewing subject. The precision machine crafted components provide for smooth movements and adjustments in positions with the knurled rotation knobs. The travel on the XY Base is as follows: X-axis allows for 1.5″ and the Y axis allows for 1.25″ of movement. Travel Area: X:1.5″ / Y:1.25″ X/Y Table Size: 3.25" x 4.25" It can be conveniently attached to compatible stands with the use of C-Clamps or the MS15X-P4 Clamp that are included in the packaging. Compatible stands: RK-10A, RK-06A, MS35B, MS37B, MS45BX-D2
Catalog No. NC1212662 Supplier Dino Lite Supplier No. MS15XS2
MS15X-S1 is a multifunctional X/Y fine adjustment base designed for precise adjustment under Dino-Lite microscopes. The included rotating table connects directly to the top plate of the X/Y stage, allowing for simple rotation of the viewing subject. The precision machine crafted components provide for smooth movements and adjustments in positions with the knurled rotation knobs. The travel on the XY Base is as follows: X-axis allows for 1.5″ and the Y axis allows for 1.25″ of movement. Travel Area: X:1.5″ / Y:1.25″ X/Y Table Size: 3.25" x 4.25" It can be conveniently attached to compatible stands with the use of C-Clamps or the MS15X-P4 Clamp that are included in the packaging. Compatible stands: RK-10A, RK-06A, MS35B, MS37B, MS45BX-D2