Learn More
Medchemexpress LLC HY-120210 5mg Medchemexpress, XY018 CAS:1873358-87-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1202105MG
Encompass_Preferred
Medchemexpress, HY-120210 5mg XY018 CAS:1873358-87-2 XY018 is a potent ROR-γ-selective antagonist. XY018 inhibits ROR-γ constitutive activity in 293T cells with high potency (EC50, 190 nM). XY018 binds to the ROR-γ hydrophobic ligand binding domain (LBD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9871