Learn More
Medchemexpress LLC XY028-133 | 2229974-73-4 | 98.6% | 25 MG
SDP
Catalog No. 5000342754
Encompass_Preferred
XY028-133 is a PROTAC-based degrader targeting CDK4/6, exhibiting anti-tumor activity by consisting of ligands for von Hippel-Lindau and cyclin-dependent kinase. It significantly reduces levels of key cell cycle proteins.
  • PROTAC-based degrader
  • Exhibits anti-tumor activity
  • Consists of ligands for von Hippel-Lindau and cyclin-dependent kinase
  • Decreases CDK4/6 protein levels
  • Decreases Cyclin A, PLK1, and pRb protein levels
Catalog No. 50-003-42754 Supplier Medchemexpress LLC Supplier No. 25MGHY129180
XY028-133 is a PROTAC-based degrader targeting CDK4/6, exhibiting anti-tumor activity by consisting of ligands for von Hippel-Lindau and cyclin-dependent kinase. It significantly reduces levels of key cell cycle proteins.
  • PROTAC-based degrader
  • Exhibits anti-tumor activity
  • Consists of ligands for von Hippel-Lindau and cyclin-dependent kinase
  • Decreases CDK4/6 protein levels
  • Decreases Cyclin A, PLK1, and pRb protein levels