Learn More
Medchemexpress LLC XY028-133 PROTAC-based CDK4/6 degrader | 2229974-73-4 | 98.62% | 1 ML
SDP
Catalog No. 5000341361
Encompass_Preferred
XY028-133 is a PROTAC-based CDK4/6 degrader exhibiting anti-tumor activity. It is composed of ligands for von Hippel-Lindau and CDK, and has been shown to significantly reduce protein levels of CDK4/6, Cyclin A, PLK1, and pRb in A375 cells at concentrations of 1 and 3 μM over 24 hours.
  • PROTAC-based CDK4/6 degrader
  • Exhibits anti-tumor activity
  • Composed of ligands for von Hippel-Lindau and CDK
  • Reduces protein levels of CDK4/6, Cyclin A, PLK1, and pRb
  • Targets CDK4 and CDK6
Catalog No. 50-003-41361 Supplier Medchemexpress LLC Supplier No. HY12918010MM/1ML
XY028-133 is a PROTAC-based CDK4/6 degrader exhibiting anti-tumor activity. It is composed of ligands for von Hippel-Lindau and CDK, and has been shown to significantly reduce protein levels of CDK4/6, Cyclin A, PLK1, and pRb in A375 cells at concentrations of 1 and 3 μM over 24 hours.
  • PROTAC-based CDK4/6 degrader
  • Exhibits anti-tumor activity
  • Composed of ligands for von Hippel-Lindau and CDK
  • Reduces protein levels of CDK4/6, Cyclin A, PLK1, and pRb
  • Targets CDK4 and CDK6