Learn More
Medchemexpress LLC HY-138946 5mg , XY028-140 CAS:2229974-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1389465MG
Encompass_Preferred
HY-138946 5mg XY028-140 CAS: 2229974-83-6 XY028-140 is a PROTAC connected by ligands for Cereblon and CDK. XY028-140 inhibits both CDK4/6 expression and CDK4/6 activity in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0818