Learn More
TARGETMOL CHEMICALS INC XY101 5MG
SDP
Catalog No. 502581357
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XY101 is a selective metabolically stable and orally available agonist of ROR(gamma) inverse(IC50 of 30 nM and a Kd of 380 nM). purity: 98%
Catalog No. 50-258-1357 Supplier TARGETMOL CHEMICALS INC Supplier No. T133625MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. XY101 is a selective metabolically stable and orally available agonist of ROR(gamma) inverse(IC50 of 30 nM and a Kd of 380 nM). purity: 98%