Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-Xyl-Sdp(O)

Catalog No. 79871150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-871-150MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-871-150MG Supplier Sigma Aldrich Supplier No. 79871150MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chiral ligand introduced in collaboration with Prof. Steve Zhou and coworkers, has shown utility in asymmetric Heck reactions for both intra and intermolecular C-C bond formation with various triflates and halides.Palladium-Catalyzed Asymmetric Intermolecular Cyclization

Specifications

CAS 1810004-87-5
Molecular Formula C20H21NO4
Linear Formula C20H21NO4
Quantity 100 mg
Synonym Amino (PEG)2 benzophenone alkyne; Probe building block
Molecular Weight (g/mol) 339.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.