Learn More
Medchemexpress LLC Xylan | 9014-63-5 | MFCD00082148 | 95.0% | 574.53 g/mol | (C5H8O4)n | 500 MG
SDP
Catalog No. 5000242077
Encompass_Preferred
Xylan is a plant-derived hemicellulose polysaccharide composed of β-1,4-linked xylose units. Supplied as a white to off-white powder (95.0% purity), CAS 9014-63-5, with a representative molecular weight of ~574.5 g/mol, this material is provided in a 500 mg pack for research and analytical applications.
  • Polymeric hemicellulose with repeating unit (C5H8O4)n.
  • Reported purity of 95.0% suitable for research use.
  • Solid powder form for easy handling and storage.
  • Provided as a 500 mg pack for small-scale experiments.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Used in biochemical and analytical studies of plant polysaccharides.
Catalog No. 50-002-42077 Supplier Medchemexpress LLC Supplier No. HY107846500MG
Xylan is a plant-derived hemicellulose polysaccharide composed of β-1,4-linked xylose units. Supplied as a white to off-white powder (95.0% purity), CAS 9014-63-5, with a representative molecular weight of ~574.5 g/mol, this material is provided in a 500 mg pack for research and analytical applications.
  • Polymeric hemicellulose with repeating unit (C5H8O4)n.
  • Reported purity of 95.0% suitable for research use.
  • Solid powder form for easy handling and storage.
  • Provided as a 500 mg pack for small-scale experiments.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Used in biochemical and analytical studies of plant polysaccharides.