Learn More
Sigma Aldrich Fine Chemicals Biosciences Xylanase powder 2500 units50G
Catalog No. 502903184
Encompass
Xylanasenbspcatalyses the degradation of xylan into xylooligosaccharides and D-xylose. It also breaks down hemicellulose. Xylanasenbspis used for the production of biofuels such as ethanol and xylitol. It improves the quality of bread and juices. Xylanase is used in biobleaching process and decreases environmental pollution.
Catalog No. 50-290-3184 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. X275350G
Xylanasenbspcatalyses the degradation of xylan into xylooligosaccharides and D-xylose. It also breaks down hemicellulose. Xylanasenbspis used for the production of biofuels such as ethanol and xylitol. It improves the quality of bread and juices. Xylanase is used in biobleaching process and decreases environmental pollution.