Learn More
Apexbio Technology LLC Xylazine HCl, 100mg. Cas: 23076-35-9 MFCD: MFCD00058196. α2 Adrenoceptor agonist
SDP
Supplier:  Apexbio Technology LLC B1351100
Encompass_Preferred
Xylazine HCl is a strong agonist of 2-adrenoceptor [1]. Xylazine HCl has been reported to stimulate the proliferation of rat thymocytes in vivo (at doses of 2.5 mg/kg and 5 mg/kg) and in vitro (at concentration of 5 mM) [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5220