Learn More
Bachem Xylazine · HCl
SDP
Supplier:  Bachem 4039613.0025
Encompass_Preferred
Xylazine · HCl, 23076-35-9, C12H16N2S · HCl, Mr 256.80, 25g. (5,6-Dihydro-4H-[1,3]thiazin-2-yl)-(2,6-dimethyl-phenyl)-amine · HCl, N-(2,6-Dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine · HCl. Xylazine is used in veterinary medicine as an analgesic, sedative, and muscle relaxant.
Catalog No. 50-260-625