Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylazine hydrochloride, MP Biomedicals™

Catalog No. MP215830701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215830701 1 g
MP215830705 5 g
MP215830725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215830701 Supplier MP Biomedicals, Inc Supplier No. 0215830701
Add to Cart
Add to Cart

Xylazine hydrochloride, White Powder

  • It has been used together to produce rapid and reversible anesthesia in experimental animals.
  • Xylazine elicited a diuretic response in ketamine-anesthesized animals, probably through activation of complex peripheral and CNS α2-adrenergic receptor systems.
  • It is an alpha agonist with sedative, analgesic and muscle relaxant properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00058196
Synonym 2-[2,6-Dimethylphenylamino]-4H-5,6-dihydroxthiazine, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride

Specifications

Melting Point 161°C to 171°C
Color White
Loss on Drying 0.0039
Molecular Formula C12H16N2S . HCl
Quantity 1 g
UN Number UN2811
Beilstein 6448471
Solubility Information Soluble in water (50mg/mL).
Physical Form Powder
Assay 1.0018
RTECSNumber XJ0776350
storageNote1 Store at Room Temperature(15-30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.