Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. X005925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X005925G 25 g
X00595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X005925G Supplier TCI America Supplier No. X005925G

Chemical Identifiers

CAS 23076-35-9
Molecular Formula C12H17ClN2S
Molecular Weight (g/mol) 256.79
MDL Number MFCD00058196
InChI Key QYEFBJRXKKSABU-UHFFFAOYSA-N
Synonym xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
PubChem CID 68554
IUPAC Name hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
SMILES [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1

Specifications

Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 256.79
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Xylazine Hydrochloride
RTECSNumber XJ0776350
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.