Learn More
BTNX Inc Xylazine Test Strip - 10pk (Liquid/Powder)
Catalog No. NC3361237
Encompass_Preferred
The Rapid Response™ Xylazine Test Strip (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in suspicious substances (liquids or powders) at the cut-off concentration of 1000 ng/mL. For forensic use only. Not an IVD.
Catalog No. NC3361237 Supplier BTNX Inc Supplier No. XYL18S210
The Rapid Response™ Xylazine Test Strip (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in suspicious substances (liquids or powders) at the cut-off concentration of 1000 ng/mL. For forensic use only. Not an IVD.