Learn More
BTNX Inc Xylazine Test Strip Kit -5pk w/ accesorries (Liquid/Powder)
Catalog No. NC3361236
Encompass_Preferred
The Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in suspicious substances (liquids or powders) at the cut-off concentration of 1000 ng/mL. For forensic use only. Not an IVD.
Catalog No. NC3361236 Supplier BTNX Inc Supplier No. XYL18S25K
The Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in suspicious substances (liquids or powders) at the cut-off concentration of 1000 ng/mL. For forensic use only. Not an IVD.