Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Xylene
SDP

Catalog No. STL84001
Change view
Click to view available options
Product Type:
Clearing Agent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
STL84001 Clearing Agent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. STL84001 Supplier StatLab Supplier No. 84001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Purified and filtered Xylene to deliver a high quality, consistent product.

  • Purified and filtered to deliver a high quality, consistent product
  • High solvency factor ensures quick and complete clearing
  • Free of impurities and water
  • Gallon Size

Specifications

Product Type Clearing Agent
Quantity 4 x 1 gal.
For Use With (Application) Tissue
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.